missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ANGPTL3 Recombinant Rabbit Monoclonal Antibody (23GB4320), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | ANGPTL3 |
|---|---|
| Clone | 23GB4320 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088172
![]() |
Invitrogen™
MA552684 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089783
|
Invitrogen™
MA552685 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ANGPTL3 Recombinant Monoclonal antibody specifically detects ANGPTL3 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| ANGPTL3 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q9Y5C1 | |
| 27329 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB4320 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| ANG-5; angiopoietin 5; angiopoietin like 3; Angiopoietin-5; angiopoietin-like 3; angiopoietin-like protein 3; Angiopoietin-related protein 3; ANGPT5; ANGPTL3; ANGPTL3(17-221); ANGPTL3(17-224); ANL3; FHBL2; hypl; hypolipidemia; UNQ153/PRO179 | |
| A synthesized peptide derived from human ANGPTL3 (17-93AA). | |
| Primary | |
| -20°C | |
| ANGPTL3 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title