missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ANGPTL3 Recombinant Rabbit Monoclonal Antibody (23GB4320), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552685
This item is not returnable.
View return policy
Description
This gene encodes a member of a family of secreted proteins that function in angiogenesis. The encoded protein, which is expressed predominantly in the liver, is further processed into an N-terminal coiled-coil domain-containing chain and a C-terminal fibrinogen chain. The N-terminal chain is important for lipid metabolism, while the C-terminal chain may be involved in angiogenesis. Mutations in this gene cause familial hypobetalipoproteinemia type 2.
Specifications
| ANGPTL3 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| ANGPTL3 | |
| ANG-5; angiopoietin 5; angiopoietin like 3; Angiopoietin-5; angiopoietin-like 3; angiopoietin-like protein 3; Angiopoietin-related protein 3; ANGPT5; ANGPTL3; ANGPTL3(17-221); ANGPTL3(17-224); ANL3; FHBL2; hypl; hypolipidemia; UNQ153/PRO179 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 27329 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB4320 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9Y5C1 | |
| ANGPTL3 | |
| A synthesized peptide derived from human ANGPTL3 (17-93AA). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction