missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Angiopoietin-like Protein 4/ANGPTL4 Antibody (Nanyang Tech.U. patent anti-ANGPTL4) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
£162.00 - £695.00
Specifications
| Antigen | Angiopoietin-like Protein 4/ANGPTL4 |
|---|---|
| Clone | Nanyang Tech.U. patent anti-ANGPTL4 |
| Applications | Flow Cytometry, ELISA, Functional Assay |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30045782
|
Novus Biologicals™
NBP3-28427-100ug |
100 μg |
£257.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30045257
|
Novus Biologicals™
NBP3-28427-1mg |
1 mg |
£695.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30045847
|
Novus Biologicals™
NBP3-28427-50ug |
50 μg |
£162.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Angiopoietin-like Protein 4/ANGPTL4 Monoclonal antibody specifically detects Angiopoietin-like Protein 4/ANGPTL4 in Human samples. It is validated for Flow Cytometry,ELISA,Functional AssaySpecifications
| Angiopoietin-like Protein 4/ANGPTL4 | |
| Flow Cytometry, ELISA, Functional Assay | |
| Unconjugated | |
| Human | |
| Apoptosis, Cancer, Hypoxia, Lipid and Metabolism, Transcription Factors and Regulators | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| angiopoietin-like 4, Angiopoietin-like protein 4, ARP4fasting-induced adipose factor, FIAFhepatic angiopoietin-related protein, Hepatic fibrinogen/angiopoietin-related protein, HFARPANGPTL2, NL2, peroxisome proliferator-activated receptor (PPAR) gamma inducedangiopoietin-related protein, PGARangiopoietin-related protein 4, pp1158, PPARG angiopoietin related protein | |
| ANGPTL4 | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. |
| Nanyang Tech.U. patent anti-ANGPTL4 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Q9BY76 | |
| 51129 | |
| IgG1 | |
| Protein A purified | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title