missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Angiopoietin-like Protein 4/ANGPTL4 Antibody (Nanyang Tech.U. patent anti-ANGPTL4) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28427-50ug
This item is not returnable.
View return policy
Description
Angiopoietin-like Protein 4/ANGPTL4 Monoclonal antibody specifically detects Angiopoietin-like Protein 4/ANGPTL4 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Angiopoietin-like Protein 4/ANGPTL4 | |
| Monoclonal | |
| Unconjugated | |
| Q9BY76 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| Nanyang Tech.U. patent anti-ANGPTL4 | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| angiopoietin-like 4, Angiopoietin-like protein 4, ARP4fasting-induced adipose factor, FIAFhepatic angiopoietin-related protein, Hepatic fibrinogen/angiopoietin-related protein, HFARPANGPTL2, NL2, peroxisome proliferator-activated receptor (PPAR) gamma inducedangiopoietin-related protein, PGARangiopoietin-related protein 4, pp1158, PPARG angiopoietin related protein | |
| ANGPTL4 | |
| 50 μg | |
| Apoptosis, Cancer, Hypoxia, Lipid and Metabolism, Transcription Factors and Regulators | |
| 51129 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction