missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Angiogenin Antibody (MM0044-7M15) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£159.00 - £423.00
Specifications
| Antigen | Angiogenin |
|---|---|
| Clone | MM0044-7M15 |
| Concentration | LYOPH |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18216732
|
Novus Biologicals
NB110-93512-0.025MG |
0.025 mg |
£159.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18246740
|
Novus Biologicals
NB110-93512 |
0.1 mg | |||||||
Description
Angiogenin Monoclonal specifically detects Angiogenin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Angiogenin | |
| LYOPH | |
| Unconjugated | |
| Mouse | |
| Angiogenesis, Cancer | |
| P03950 | |
| 283 | |
| human recombinant Angiogenin protein | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| MM0044-7M15 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| ALS9, angiogenin, angiogenin, ribonuclease, RNase A family, 5, EC 3.1.27, EC 3.1.27.-, epididymis luminal protein 168, HEL168, MGC22466, MGC71966, Ribonuclease 5, RNase 5, RNASE5RNASE4 | |
| ANG | |
| IgG1 | |
| Protein G purified | |
| This recognizes human Angiogenin in western blot. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title