missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Angiogenin Antibody (MM0044-7M15) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-93512
This item is not returnable.
View return policy
Description
Angiogenin Monoclonal specifically detects Angiogenin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Angiogenin | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:100-1:1000, Immunohistochemistry 1:25-1:50, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:25-1:50 | |
| P03950 | |
| ANG | |
| human recombinant Angiogenin protein | |
| 0.1 mg | |
| Angiogenesis, Cancer | |
| 283 | |
| Reconstitute with sterilized PBS to a final concentration of 0.2 mg/ml. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| MM0044-7M15 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS. 0.025 mg size is provided in liquid form, PBS with No Preservative | |
| ALS9, angiogenin, angiogenin, ribonuclease, RNase A family, 5, EC 3.1.27, EC 3.1.27.-, epididymis luminal protein 168, HEL168, MGC22466, MGC71966, Ribonuclease 5, RNase 5, RNASE5RNASE4 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| This recognizes human Angiogenin in western blot. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction