Learn More
ABEBA™ Anatom Green/Yellow Safety Sandale S1P ESD
Green/yellow sandals made of water/tear resistant Cordura, slip resistant PU2D outsole, steel toe cap, textile antipuncture
Brand: ABEBA™ 32283/49
Additional Details : Weight : 1.41000kg
Description
- Comfortable, robust, durable sandals
- Comply to EN ISO 20345: 2012, S1P, SRC
- ESD according EN 61340 class climatic 1
- Slip resistant PU double density outsole
- Green/yellow water and tear resistant Cordura
- Steel toe cap (200 J)
- Textile antipuncture midsole
- Scratch-resistant coated fabric in toe and heel area
- Breathable and antibacterial lining with silver ions
- Removable active comfort insole
Specifications
Green, Yellow | |
49 | |
EN ISO 20345: 2012 S1P SRC, EN 61340 ESD class climatic 1 | |
Sandals | |
S1P SRC ESD |
Cordura | |
Steel | |
Men's | |
No |