Learn More
IKA™ Analytical Batch Mill
Grinds hard, brittle, soft and fibrous materials. IKA™ Analytical Batch Mill allows samples to be embrittled directly in the grinding chamber, tough, oily and aqueous samples can also be ground. Available models include the newly redesigned A 10 Basic and the A 11 Basic.
POA
Specifications
Product Type | Analytical Batch Mill |
---|---|
Hertz | 50/60 Hz |
Material | Stainless Steel |
Description
Grinds hard, brittle, soft and fibrous materials. IKA™ Analytical Batch Mill allows samples to be embrittled directly in the grinding chamber, tough, oily and aqueous samples can also be ground. Available models include the newly redesigned A 10 Basic and the A 11 Basic.
Grinds hard, brittle, soft and fibrous materials Samples may be embrittled directly in the grinding chamber, tough, oily and aqueous samples can also be ground.A 10 Basic: Redesigned batch mill grinds hard, brittle, soft and fibrous materials for volumes up to 50mL. The development of the mill placed particular emphasis on safety. The mill will only start when the lid is closed and can only be opened at standstill. A quick stop feature further increases the safety of the user.
- Digital timer
- Counter display of grinding time
- Interval function
- Brushless motor for long service life and low noise level Integrated cooling connections
- Error code display
- Simple handling with keypads
- Easily exchangeable beater/cutter for a variety of applications
- Grinding chamber reduction for small sample sizes and stainless steel cutter A 10.1 included with delivery
- Bayonet lock for lid to enable simple and safe locking
- Moist and gluey materials can be pulverized by adding water
- Grinding chamber made of Tefcel (PTFE, glass fiber-reinforced) with stainless steel inlet (1.4571), useful volume 80mL (incl. with delivery). For embrittlement of grinding materials with liquid nitrogen in the grinding chamber
Specifications
Analytical Batch Mill | |
50/60 Hz | |
Stainless Steel |