missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Anaerobe Blood Agar (WC) with Nalidixic Acid + Tween

Product Code. 12917128
Click to view available options
Description:
Anaerobe Blood (WC) Nalidixic Acid + Tween
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 12917128

Brand: Thermo Scientific™ PB0113A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Isolate, cultivate and/ or undertake susceptibility testing of non-sporing anaerobes with this selective, ready-to-use medium.

Isolate, cultivate non-sporing anaerobes and/or undertake susceptibility testing of aerobic bacteria from clinical samples using Thermo Scientific™ Oxoid™ Anaerobe Blood Agar (WC) with Nalidixic acid + TWEEN™ commonly known as NAT medium. The presence of Nalidixic acid makes the medium selective for non-sporing anaerobes, while TWEEN 80 stimulates the growth of Gram positive anaerobes and slowly lyses the blood allowing early colonial pigmentation by Prevotella Melaninogenica1.

Use Anaerobe Blood Agar (WC) with Nalidixic acid + TWEEN for the isolation of non-sporing anaerobes from clinical samples and for antimicrobial susceptibility testing of aerobic bacteria using the Receiver Operating Characteristic (ROC) procedure. The medium utilizes Thermo Scientific™Oxoid™ Wilkins Chalgren Agar as a base2.

  • Selective for non-sporing anaerobes: presence of Nalidixic acid and TWEEN makes it selective for non-sporing anaerobes.
  • Superior performance: superior recovery of non-clostridial anaerobes compared to that of kanamycin and neomycin agars3.
  • Ready-to-use convenience of prepared media

The recovery of non-sporing anaerobes from clinical material may sometimes prove difficult in specimens containing mixture of aerobic and anaerobic bacteria. As described by Wren4, the presence of Nalidixic acid makes the medium virtually non-inhibitory to most non-sporing anaerobes While retaining good selectivity for these organisms when present in mixed cultures. The medium is patiularly useful for the recovery of non-sporing Gram positive anaerobes since the presence of TWEEN 80 stimulates their growth.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Color Ruby Red
Content And Storage 2°C to 10°C
Description Anaerobe Blood (WC) Nalidixic Acid + Tween
Format 90mm Monoplate
Manufacturing Origin United Kingdom
Product Type Agar
Fill Weight 19.5g±1.0.g
Packaging Type 10 x 90mm plates, film wrapped
Quantity 10/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.