missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMSH-LP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-56334
This item is not returnable.
View return policy
Description
AMSH-LP Polyclonal specifically detects AMSH-LP in Human samples. It is validated for Western Blot.
Specifications
| AMSH-LP | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| ALMalpha, AMSH-FP, AMSHLP, AMSH-LPassociated molecule with the SH3 domain of STAM (AMSH) - Family Protein, associated molecule with the SH3 domain of STAM (AMSH) like protein, bA399O19.2, EC 3.1.2.15, EC 3.4.19.-, FLJ31524, KIAA1373AMSH-like protease, STAM binding protein-like 1, STAM-binding protein-like 1 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Expected identity based on immunogen sequence: Canine: 100%; Equine: 100%; Mouse: 100%; Pig: 100%; Xenopus: 92%; Bovine: 92%; Chicken: 78%. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q96FJ0 | |
| STAMBPL1 | |
| Synthetic peptides corresponding to STAMBPL1(STAM binding protein-like 1) The peptide sequence was selected from the N terminal of STAMBPL1. Peptide sequence MDQPFTVNSLKKLAAMPDHTDVSLSPEERVRALSKLGCNITISEDITPRR. | |
| 100 μL | |
| DNA replication Transcription Translation and Splicing | |
| 57559 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction