missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Amrinone, 98%

Phosphodiesterase III inhibitor and inhibits platelet aggregation

£56.30 - £162.00

Chemical Identifiers

CAS 60719-84-8
Formula Weight 187.20g/mol
Synonym 5-Amino-(3,4′-bipyridin)-6(1H)-one
Content And Storage 2°C to 8°C
Format Crystals
For Use With (Application) Phosphodiesterase III inhibitor and inhibits platelet aggregation
Description Yellow
Melting Point 294°C to 297°C (dec.)
Solubility Insoluble in water or chloroform; Slightly soluble in acetic acid or DMF; Soluble in DMSO
Assay 98%
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15404989
SDS View Documents
Thermo Scientific™
J62655.MD
250 mg
ONLINE EXCLUSIVE PRICE
£57.50
£56.30 / 250mg
Save £1.20
2% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15494979
SDS View Documents
Thermo Scientific™
J62655.03
ONLINE EXCLUSIVE PRICE
£170.00
£162.00 / 1g
Save £8.00
4% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

60719-84-8
5-Amino-(3,4′-bipyridin)-6(1H)-one
Crystals
Yellow
Insoluble in water or chloroform; Slightly soluble in acetic acid or DMF; Soluble in DMSO
187.20g/mol
2°C to 8°C
Phosphodiesterase III inhibitor and inhibits platelet aggregation
294°C to 297°C (dec.)
98%

Specifications

Yellow
Crystals
60719-84-8
187.20g/mol
294°C to 297°C (dec.)
98%
2°C to 8°C
Phosphodiesterase III inhibitor and inhibits platelet aggregation
C10H9N3O
5-Amino-(3,4′-bipyridin)-6(1H)-one
Insoluble in water or chloroform; Slightly soluble in acetic acid or DMF; Soluble in DMSO
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 262-390-0

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.