Learn More
Thermo Scientific™ Amrinone, 98%
Phosphodiesterase III inhibitor and inhibits platelet aggregation
Brand: Thermo Scientific™ J62655.MD
| Quantity | 250 mg |
|---|
View more versions of this product
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 60719-84-8 | |
| 5-Amino-(3,4′-bipyridin)-6(1H)-one | |
| Crystals | |
| Yellow | |
| Insoluble in water or chloroform; Slightly soluble in acetic acid or DMF; Soluble in DMSO |
| 187.20g/mol | |
| 2°C to 8°C | |
| Phosphodiesterase III inhibitor and inhibits platelet aggregation | |
| 294°C to 297°C (dec.) | |
| 98% |
Specifications
| 250 mg | |
| 2°C to 8°C | |
| Phosphodiesterase III inhibitor and inhibits platelet aggregation | |
| C10H9N3O | |
| 5-Amino-(3,4′-bipyridin)-6(1H)-one | |
| Insoluble in water or chloroform; Slightly soluble in acetic acid or DMF; Soluble in DMSO |
| Yellow | |
| Crystals | |
| 60719-84-8 | |
| 187.20g/mol | |
| 294°C to 297°C (dec.) | |
| 98% |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.