missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Phosphate GFAA Matrix Modifier: 10% NH4H2PO4 in 2% HNO3, VHG™

Product Code. 17903626
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17903626 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17903626 Supplier VHG Labs Supplier No. VHGMAP10P100
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Ammonium Phosphate GFAA Matrix Modifier: 10% NH4H2PO4 in 2% HNO3
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ammonium Phosphate
CAS 10361-65-6
Methods GFAA
Analyte or Component Names Ammonium Phosphate
Shipping Condition Room Temperature
Molecular Formula (NH4)3PO4
Concentration 10%
Synonym (NH4)3PO4
SMILES [NH4+].[NH4+].[NH4+].[O-]P(=O)([O-])[O-]
IUPAC Name Ammonium Phosphate
Solvent or Matrix Dilute HNO3
Molecular Weight (g/mol) 149.09
Quantity 100 mL
Formula Weight 149.09 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.