Learn More
Ammonium hexachlororhodate(III) hydrate, Premion™, 99.995% (metals basis), Thermo Scientific Chemicals
POA - £2679.00
Chemical Identifiers
CAS | 15336-18-2 |
---|---|
Molecular Formula | Cl6H12N3Rh |
Molecular Weight (g/mol) | 369.72 |
MDL Number | MFCD00003394 |
InChI Key | QNMMDVLCBKRUJU-UHFFFAOYSA-K |
Synonym | ammonium hexachlororhodate iii, triammonium hexachlororhodate, triammonium hexachlororhodate 3-, epitope id:151523, rhodium iii-ammonium chloride, ammonium hexachloridorhodate 3-, ammonium hexachloridorhodate iii, ammonium hexachlororhodate iii , reagent grade |
PubChem CID | 16211509 |
ChEBI | CHEBI:60128 |
SMILES | [NH4+].[NH4+].[NH4+].Cl[Rh+7](Cl)(Cl)(Cl)(Cl)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11346997
|
Thermo Scientific Alfa Aesar
010849.02 |
0.5 g | |||||||
11356997
|
Thermo Scientific Alfa Aesar
010849.03 |
1 g |
£624.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11336997
|
Thermo Scientific Alfa Aesar
010849.06 |
5 g |
£2679.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is used as an intermediate in organic synthesis for the preparation of pharmaceuticals and agrochemicals. It can be used as a precursor for the functionalization of multi-walled carbon nanotubes (MWCNT) with metallic nanoparticles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
15336-18-2 | |
369.72 | |
QNMMDVLCBKRUJU-UHFFFAOYSA-K | |
16211509 | |
[NH4+].[NH4+].[NH4+].Cl[Rh+7](Cl)(Cl)(Cl)(Cl)Cl |
Cl6H12N3Rh | |
MFCD00003394 | |
ammonium hexachlororhodate iii, triammonium hexachlororhodate, triammonium hexachlororhodate 3-, epitope id:151523, rhodium iii-ammonium chloride, ammonium hexachloridorhodate 3-, ammonium hexachloridorhodate iii, ammonium hexachlororhodate iii , reagent grade | |
CHEBI:60128 |
Specifications
(decomposition) | |
Red | |
Cl6H12N3Rh | |
ammonium hexachlororhodate iii, triammonium hexachlororhodate, triammonium hexachlororhodate 3-, epitope id:151523, rhodium iii-ammonium chloride, ammonium hexachloridorhodate 3-, ammonium hexachloridorhodate iii, ammonium hexachlororhodate iii , reagent grade | |
QNMMDVLCBKRUJU-UHFFFAOYSA-K | |
triazanium;hexachlororhodium(3-) | |
16211509 | |
369.74 (Anhydrous) | |
Premion™ | |
(metals basis) |
15336-18-2 | |
Powder | |
MFCD00003394 | |
Soluble in water | |
[NH4+].[NH4+].[NH4+].Cl[Rh+7](Cl)(Cl)(Cl)(Cl)Cl | |
369.72 | |
CHEBI:60128 | |
99.995% | |
Hygroscopic | |
Ammonium hexachlororhodate(III) hydrate |
Safety and Handling
GHS H Statement
H318
Causes serious eye damage.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
missing translation for 'einecsNumber' : 239-364-2
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only