missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hexachlororhodate(III) hydrate, Premion™, 99.995% (metals basis)

Product Code. 11356997
Change view
Click to view available options
Quantity:
0.5 g
1 g
5 g
Unit Size:
0.5g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11356997 1 g 1g
11346997 0.5 g 0.5g
11336997 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11356997 Supplier Thermo Scientific Alfa Aesar Supplier No. 010849.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 15336-18-2 | Cl6H12N3Rh | 369.72 g/mol

It is used as an intermediate in organic synthesis for the preparation of pharmaceuticals and agrochemicals. It can be used as a precursor for the functionalization of multi-walled carbon nanotubes (MWCNT) with metallic nanoparticles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15336-18-2
Molecular Formula Cl6H12N3Rh
Molecular Weight (g/mol) 369.72
MDL Number MFCD00003394
InChI Key QNMMDVLCBKRUJU-UHFFFAOYSA-K
Synonym ammonium hexachlororhodate iii, triammonium hexachlororhodate, triammonium hexachlororhodate 3-, epitope id:151523, rhodium iii-ammonium chloride, ammonium hexachloridorhodate 3-, ammonium hexachloridorhodate iii, ammonium hexachlororhodate iii , reagent grade
PubChem CID 16211509
ChEBI CHEBI:60128
SMILES [NH4+].[NH4+].[NH4+].Cl[Rh+7](Cl)(Cl)(Cl)(Cl)Cl

Specifications

Melting Point (decomposition)
Color Red
Physical Form Powder
Quantity 1 g
Sensitivity Hygroscopic
Solubility Information Soluble in water
IUPAC Name triazanium;hexachlororhodium(3-)
Formula Weight 369.74 (Anhydrous)
Percent Purity 99.995%
Grade Premion
Assay (metals basis)
Chemical Name or Material Ammonium hexachlororhodate(III) hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.