missing translation for 'onlineSavingsMsg'
Learn More

Ammonium fluoride hydrate, Puratronic™, 99.995% (metals basis)

Product Code. 11359367
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11359367 25 g 25g
11349367 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11359367 Supplier Thermo Scientific Alfa Aesar Supplier No. 040216.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12449-35-3 | FH4N | 37.04 g/mol

Ammonium fluoride is used for etching of glass, preservation of food, in printing textile, and as mothproofing agent. It is used in chemical analysis and in brewing. It is used in the synthesis of NaHoF4 and NaEuF4, which are important in solid state lasers as well as photoluminescence. The fluoride ion is used as a base and nucleophile in organic synthesis. Ammonium fluoride providee safe deprotection strategy for the tert-butyldimethylsilyl (TBS) group during RNA synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12449-35-3
Molecular Formula FH4N
Molecular Weight (g/mol) 37.04
MDL Number MFCD00011423
InChI Key LDDQLRUQCUTJBB-UHFFFAOYSA-N
IUPAC Name amine hydrofluoride
SMILES N.F

Specifications

Color White
Physical Form Crystalline Aggregates
Quantity 25 g
Linear Formula NH4F·xH2O (x≈0.5)
UN Number UN2505
Solubility Information Freely soluble in water. Slightly soluble in alcohol.
Formula Weight 37.04 (Anhydrous)
Percent Purity 99.9975%
Grade Puratronic
Odor Odorless
Assay (metals basis)
Chemical Name or Material Ammonium fluoride hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.