CAS RN 12449-35-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Ammonium fluoride hydrate, Puratronic™, 99.995% (metals basis)
CAS: 12449-35-3 Molecular Formula: FH4N Molecular Weight (g/mol): 37.04 MDL Number: MFCD00011423 InChI Key: LDDQLRUQCUTJBB-UHFFFAOYSA-N IUPAC Name: amine hydrofluoride SMILES: N.F