missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Acetate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
High quality chemicals for laboratory and specialized industrial use
£358.00
Chemical Identifiers
CAS | 631-61-8 |
---|---|
Molecular Formula | C2H7NO2 |
Molecular Weight (g/mol) | 77.083 |
InChI Key | USFZMSVCRYTOJT-UHFFFAOYSA-N |
Synonym | ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4 |
PubChem CID | 517165 |
ChEBI | CHEBI:62947 |
IUPAC Name | azanium;acetate |
SMILES | CC(=O)[O-].[NH4+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15507854
|
Avantor J.T.Baker
0596-01 |
500 g | Poly Bottle | ||||||
Chemical Identifiers
631-61-8 | |
77.083 | |
ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4 | |
CHEBI:62947 | |
CC(=O)[O-].[NH4+] |
C2H7NO2 | |
USFZMSVCRYTOJT-UHFFFAOYSA-N | |
517165 | |
azanium;acetate |
Specifications
631-61-8 | |
C2H7NO2 | |
USFZMSVCRYTOJT-UHFFFAOYSA-N | |
azanium;acetate | |
517165 | |
77.08 | |
1L = 1.07kg |
Crystals | |
ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4 | |
CC(=O)[O-].[NH4+] | |
77.083 | |
CHEBI:62947 | |
BAKER ANALYZED™ ACS Reagent | |
Ammonium Acetate |