missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Acetate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog Number 15507854
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
This item is not returnable. View return policy
631-61-8
C2H7NO2
77.083
USFZMSVCRYTOJT-UHFFFAOYSA-N
ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4
517165
CHEBI:62947
azanium;acetate
CC(=O)[O-].[NH4+]
This item is not returnable. View return policy
631-61-8
C2H7NO2
77.083
USFZMSVCRYTOJT-UHFFFAOYSA-N
ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4
517165
CHEBI:62947
azanium;acetate
CC(=O)[O-].[NH4+]

High quality chemicals for laboratory and specialized industrial use

Physical Form Crystals
Quantity 500 g
Formula Weight 77.08
Grade BAKER ANALYZED™ ACS Reagent
Packaging Poly Bottle
Specific Gravity 1L = 1.07kg
Chemical Name or Material Ammonium Acetate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'