Learn More
Amiodarone hydrochloride, Thermo Scientific™
A non-selective ion channel blocker; also an antiarrhythmic agent
£24.90 - £108.65
Chemical Identifiers
| CAS | 19774-82-4 |
|---|---|
| Molecular Formula | C25H30ClI2NO3 |
| Molecular Weight (g/mol) | 681.778 |
| MDL Number | MFCD00069204 |
| InChI Key | ITPDYQOUSLNIHG-UHFFFAOYSA-N |
| Synonym | amiodarone hydrochloride, amiodarone hcl, amiodar, nexterone, pacerone, amiodaronum hydrochloride, ritmocardyl, rythmarone, angoron |
| PubChem CID | 441325 |
| IUPAC Name | (2-butyl-1-benzofuran-3-yl)-[4-[2-(diethylamino)ethoxy]-3,5-diiodophenyl]methanone;hydrochloride |
| SMILES | CCCCC1=C(C2=CC=CC=C2O1)C(=O)C3=CC(=C(C(=C3)I)OCCN(CC)CC)I.Cl |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15493219
|
Thermo Scientific Alfa Aesar
J60456.03 |
1g |
£24.90
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15403229
|
Thermo Scientific Alfa Aesar
J60456.06 |
5g |
£35.70 £32.65 / 5g Save £3.05 8% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15413229
|
Thermo Scientific Alfa Aesar
J60456.14 |
25g |
£121.00 £108.65 / 25g Save £12.35 10% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsAmiodarone induces an immediate influx of Ca2+ in Saccharomyces cerevisiae, followed by mitochondrial fragmentation and cell death.
Solubility
Soluble in chloroform, methanol.
Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents, heat. Store in refrigerator.
Chemical Identifiers
| 19774-82-4 | |
| 681.778 | |
| ITPDYQOUSLNIHG-UHFFFAOYSA-N | |
| 441325 | |
| CCCCC1=C(C2=CC=CC=C2O1)C(=O)C3=CC(=C(C(=C3)I)OCCN(CC)CC)I.Cl |
| C25H30ClI2NO3 | |
| MFCD00069204 | |
| amiodarone hydrochloride, amiodarone hcl, amiodar, nexterone, pacerone, amiodaronum hydrochloride, ritmocardyl, rythmarone, angoron | |
| (2-butyl-1-benzofuran-3-yl)-[4-[2-(diethylamino)ethoxy]-3,5-diiodophenyl]methanone;hydrochloride |
Specifications
| 19774-82-4 | |
| MFCD00069204 | |
| amiodarone hydrochloride, amiodarone hcl, amiodar, nexterone, pacerone, amiodaronum hydrochloride, ritmocardyl, rythmarone, angoron | |
| ITPDYQOUSLNIHG-UHFFFAOYSA-N | |
| (2-butyl-1-benzofuran-3-yl)-[4-[2-(diethylamino)ethoxy]-3,5-diiodophenyl]methanone;hydrochloride | |
| 441325 | |
| Amiodarone hydrochloride |
| C25H30ClI2NO3 | |
| 14,482 | |
| Soluble in chloroform,methanol. | |
| CCCCC1=C(C2=CC=CC=C2O1)C(=O)C3=CC(=C(C(=C3)I)OCCN(CC)CC)I.Cl | |
| 681.778 | |
| 681.77 |
Safety and Handling
P201-P202-P260-P271-P281-P302+P352-P304+P340-P308+P313-P312-P363-P501c
H312+H332-H361d-H373
missing translation for 'einecsNumber' : 243-293-2
missing translation for 'rtecsNumber' : OB1361000
missing translation for 'tsca' : No
Recommended Storage : Keep cold
- Amiodarone hydrochloride
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Reproductive toxicity Category 2
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H361d-Suspected of damaging the unborn child.
- H373-May cause damage to organs.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
- MIXTURE LIST-Contain: Amiodarone hydrochloride
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.