missing translation for 'onlineSavingsMsg'
Learn More

Amiodarone hydrochloride, Thermo Scientific™

A non-selective ion channel blocker; also an antiarrhythmic agent

Brand:  Thermo Scientific Alfa Aesar J60456.14


Quantity 25g

 View more versions of this product

Product Code. 15413229

  • ONLINE EXCLUSIVE PRICE
    £121.00
    £108.65 / 25g
    Save £12.35
    10% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Amiodarone induces an immediate influx of Ca2+ in Saccharomyces cerevisiae, followed by mitochondrial fragmentation and cell death.

Solubility
Soluble in chloroform, methanol.

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents, heat. Store in refrigerator.
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

19774-82-4
681.778
ITPDYQOUSLNIHG-UHFFFAOYSA-N
441325
CCCCC1=C(C2=CC=CC=C2O1)C(=O)C3=CC(=C(C(=C3)I)OCCN(CC)CC)I.Cl
C25H30ClI2NO3
MFCD00069204
amiodarone hydrochloride, amiodarone hcl, amiodar, nexterone, pacerone, amiodaronum hydrochloride, ritmocardyl, rythmarone, angoron
(2-butyl-1-benzofuran-3-yl)-[4-[2-(diethylamino)ethoxy]-3,5-diiodophenyl]methanone;hydrochloride

Specifications

19774-82-4
MFCD00069204
14,482
Soluble in chloroform,methanol.
CCCCC1=C(C2=CC=CC=C2O1)C(=O)C3=CC(=C(C(=C3)I)OCCN(CC)CC)I.Cl
681.778
681.77
C25H30ClI2NO3
25g
amiodarone hydrochloride, amiodarone hcl, amiodar, nexterone, pacerone, amiodaronum hydrochloride, ritmocardyl, rythmarone, angoron
ITPDYQOUSLNIHG-UHFFFAOYSA-N
(2-butyl-1-benzofuran-3-yl)-[4-[2-(diethylamino)ethoxy]-3,5-diiodophenyl]methanone;hydrochloride
441325
Amiodarone hydrochloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Amiodarone hydrochloride

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 2
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H361d-Suspected of damaging the unborn child.
  • H373-May cause damage to organs.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

Supplemental information
  • MIXTURE LIST-Contain: Amiodarone hydrochloride

SDS Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.