missing translation for 'onlineSavingsMsg'
Learn More

Aminopeptidase P1/XPNPEP1 Mouse anti-Human, Clone: 9C7, Novus Biologicals™

Mouse Monoclonal Antibody

£234.00 - £378.00

Specifications

Antigen Aminopeptidase P1/XPNPEP1
Clone 9C7
Concentration 1 mg/mL
Dilution Western Blot 1:1000, ELISA, Immunocytochemistry/Immunofluorescence 1:100
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18230693
View Documents
Novus Biologicals
NBP2-59499-50UL
50 μL
£234.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18244095
View Documents
Novus Biologicals
NBP2-59499-100UL
100 μL
£378.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Aminopeptidase P1/XPNPEP1 Monoclonal specifically detects Aminopeptidase P1/XPNPEP1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Aminopeptidase P1/XPNPEP1
1 mg/mL
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Unconjugated
Mouse
Human
Aminoacylproline aminopeptidase, APP1, cytosolic, Cytosolic aminopeptidase P, EC 3.4.11.9, sAmp, xaa-Pro aminopeptidase 1, XPNPEPLX-prolyl aminopeptidase (aminopeptidase P)-like, X-prolyl aminopeptidase (aminopeptidase P) 1, soluble, X-prolyl aminopeptidase 1, soluble
XPNPEP1
IgG2b κ
Protein A purified
9C7
Western Blot 1:1000, ELISA, Immunocytochemistry/Immunofluorescence 1:100
Monoclonal
Purified
RUO
Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide
7511
Recombinant human XPNPEP1 (1-623aa) purified from E. coli
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.