missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aminopeptidase P1/XPNPEP1 Mouse anti-Human, Clone: 9C7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59499-100UL
This item is not returnable.
View return policy
Description
Aminopeptidase P1/XPNPEP1 Monoclonal specifically detects Aminopeptidase P1/XPNPEP1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Aminopeptidase P1/XPNPEP1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, ELISA, Immunocytochemistry/Immunofluorescence 1:100 | |
| Aminoacylproline aminopeptidase, APP1, cytosolic, Cytosolic aminopeptidase P, EC 3.4.11.9, sAmp, xaa-Pro aminopeptidase 1, XPNPEPLX-prolyl aminopeptidase (aminopeptidase P)-like, X-prolyl aminopeptidase (aminopeptidase P) 1, soluble, X-prolyl aminopeptidase 1, soluble | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 7511 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2b κ |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 9C7 | |
| Unconjugated | |
| Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
| XPNPEP1 | |
| Recombinant human XPNPEP1 (1-623aa) purified from E. coli | |
| 100 μL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction