missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ AminoLink™ Immobilization Kit

Product Code. 10259883 Shop All Thermo Scientific Products
Click to view available options
Quantity:
5 Column Kit
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 10259883

Brand: Thermo Scientific™ 44890

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Covalently immobilize proteins and antibodies by lysine residues or other primary amines (-NH2) via reductive amination with these aldehyde-activated agarose beads.

Thermo Scientific™ AminoLink Coupling Resin is crosslinked 4% beaded agarose that has been activated with aldehyde groups to enable covalent immobilization of antibodies and other proteins through primary amines.

AminoLink Coupling Resin is crosslinked 4% beaded agarose that has been activated with aldehyde groups. Proteins and other molecules with primary amines can be covalently attached (immobilized) to the resin to make chromatography columns for use in affinity purification. The aldehyde groups form stable secondary amine bonds with primary amines such as exist in the side chain of lysine (K) residues, which are generally abundant and readily accessible in proteins. Once a protein is immobilized, the prepared affinity resin can be used for a variety of batch and column affinity purification methods involving binding interactions with the immobilized protein. The resin and linkage are stable in most binding and elution conditions typically used in affinity chromatography, enabling prepared resin to be used for multiple rounds of affinity purification procedures.

Highlights:

  • AminoLink Coupling Resin – aldehyde-activated crosslinked 4% beaded agarose

  • Ideal for antibodies and other proteins – immobilize molecules via primary amines (-NH2)

  • Flexible coupling conditions – efficient (>85%) coupling over a wide range of pH (4 to 10) and buffer conditions (PBS or other non-amine buffer with or without organic solvent

  • Stable, permanent immobilization – coupling reaction results in stable, leak-resistant secondary amine bond between resin and ligand

  • Better than immobilization to CNBr-activated agarose – bond is more stable and uncharged, resulting in less nonspecific binding in affinity purification procedures

  • Versatile and reusable – prepared affinity resin is adaptable to column and batch affinity techniques and the resin is reusable for typical applications based on protein binding interactions

Includes:

Kit contains pre-packed AminoLink Columns, coupling buffer, quenching buffer, wash solution, sodium cyanoborohydride solution, and column accessories

TRUSTED_SUSTAINABILITY

Specifications

Description AminoLink Immobilization Kit, 2mL
Content And Storage Sufficient For: Immobilizing 2 to 20 mg of protein per column
• AminoLink Columns, 2mL, 5 columns
• AminoLink Coupling Buffer (pH 7.0), 250 mL
• Quenching Buffer, 50 mL
• Wash Buffer, 240 mL
• Sodium Cyanoborohydride Solution, 5M, 0.5 mL
• Column Accessories

Store at 4°C
Product Line AminoLink
Quantity 5 Column Kit
Target Antibody
Type Immobilization Kit
Stationary Phase Aldehyde
Sufficient For 5 Protein Immobilizations
Format Spin Column
Column Type Affinity, Agarose Resin
compliance-icons compliance-icons compliance-icons
Product Identifier
  • AminoLink Immobilization Kit
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H400-Very toxic to aquatic life.
  • EUH032-Contact with acids liberates very toxic gas.
  • EUH204-Contains isocyanates. May produce an allergic reaction.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P262-Do not get in eyes, on skin, or on clothing.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P331-Do NOT induce vomiting.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Sodium hydroxide

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.