Learn More
Cytiva Amersham™ CyDye™ Value Packs (mono-Reactive NHS Ester)
Use for highly accurate labeling of amine groups. Cytiva Amersham™ CyDye™ Value Packs (mono-Reactive NHS Ester) contain derivitized CyDyes that carry only one reactive group on each dye molecule for accurate labeling of amine groups, for example on antibodies, peptides, or oligonucleotides.
Brand: Cytiva PA13601
Code : GE
Additional Details : Weight : 2.85310kg
Description
- Contains derivitized CyDyes that carry only one reactive group on each dye molecule
- pH range: 3 to 10
- DMSO tolerant
- Photostable
- Example protocols included with each dye pack
Specifications
For labeling via Amine groups, CyDyes are suitable for use in applications such as Fluorescence resonance energy transfer (FRET), time resolved-Fluorescence resonance energy transfer (TR-FRET) and Fluorescence polarization (FP) | |
Cy3.5 NHS Ester | |
3 to 10 | |
Cy3.5 |
1 mg | |
Foil packs, each containing 1, 5, 10, 25 or 50mg dried dye | |
2°C to 8°C |
Safety and Handling
- Danger
Hazard Category
- Acute toxicity Category 4
- Skin sensitiser Category 1
- Respiratory sensitiser Category 1