missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Amersham™ CyDye™ Value Packs (mono-Reactive Maleimide)

For the selective labeling of free sulfhydryl groups such as cysteine residues in peptides, proteins, and oligonucleotides

Brand:  Cytiva PA15136

Code : GE

Additional Details : Weight : 0.03600kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III

 View more versions of this product

Product Code. 11545085

  • £3501.00 / 25mg
Estimated Shipment: 17-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The use of mono-reactive maleimides is particularly suited to antibody labeling since the dye molecules are introduced away from the antibody binding site, increasing the chance that functionality of the labeled molecule will be unimpaired.

  • Suitable for the labeling of antibodies
  • Example protocols included with each dye pack

Applications:

Antibody labeling

Specifications

Specifications

For labelling via thiol groups For use in steady state fluorescence, FP and FRET applications
Cy5 Maleimide
3 to 10
Soluble in cold water and hot water
Cy5
25 mg
Foil packs, each containing 1, 5 or 25mg dried dye
791.95
2°C to 8°C, protected from light. Reconstituted material may be stored for up to 2 weeks at -20°C in aliquots to avoid repeat freeze thaw cycles.
Blue
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Skin sensitiser Category 1
  • Respiratory sensitiser Category 1

SDS
Documents

Documents

Certificates
Special Offers

Special Offers