Learn More
Cytiva Amersham™ CyDye™ Value Packs (mono-Reactive Maleimide)
For the selective labeling of free sulfhydryl groups such as cysteine residues in peptides, proteins, and oligonucleotides
Brand: Cytiva PA15131
Code : GE
Additional Details : Weight : 0.00100kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III
Description
The use of mono-reactive maleimides is particularly suited to antibody labeling since the dye molecules are introduced away from the antibody binding site, increasing the chance that functionality of the labeled molecule will be unimpaired.
- Suitable for the labeling of antibodies
- Example protocols included with each dye pack
Antibody labeling
Specifications
For labelling via thiol groups For use in steady state fluorescence, FP and FRET applications | |
Cy5 Maleimide | |
3 to 10 | |
Soluble in cold water and hot water | |
Cy5 |
1 mg | |
Foil packs, each containing 1, 5 or 25mg dried dye | |
791.95 | |
2°C to 8°C, protected from light. Reconstituted material may be stored for up to 2 weeks at -20°C in aliquots to avoid repeat freeze thaw cycles. | |
Blue |
Safety and Handling
- Danger
Hazard Category
- Acute toxicity Category 3
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
Hazard Statement
- H301-Toxic if swallowed.
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P501-Dispose of contents/container to .
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P405-Store locked up.