missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Amersham™ CyDye™ Value Packs (mono-Reactive Hydrazide)

For the labeling of free carbonyl groups of glycoproteins and carbohydrates

Brand:  Cytiva PA15621

Code : GE

Additional Details : Weight : 0.03700kg

 View more versions of this product

Product Code. 10369524

  • £696.00 / 1mg
Estimated Shipment: 11-07-2024
to see stock.
Add to basket

Description

Description

Hydrazide esters provide a convenient method for the labeling of free carbonyl groups of glycoproteins and carbohydrates. For maximum convenience, stable active hydrazide esters are supplied and stored as solid material.

  • Range of bulk packs available for greatest economy
  • Example protocols included with each dye pack

Specifications

Specifications

For labelling via carbonyl groups. For use in steady state fluorescence, FP and FRET applications
Cy5.5 Hydrazide
1128.42
2°C to 8°C, protected from light. Reconstituted material may be stored for up to 2 weeks at -20°C in aliquots to avoid repeat freeze thaw cycles.
Dark Blue
1 mg
3 to 10
Soluble in cold water and hot water
Cy5.5
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Skin sensitiser Category 1
  • Respiratory sensitiser Category 1

SDS
Documents

Documents

Certificates
Special Offers

Special Offers