missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Amersham™ CyDye™ Value Packs (bis-Reactive NHS Ester)

Product Code. 11585045 Shop All Cytiva Products
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Product Type:
Cy2 bis-NHS Ester
Cy3 bis-NHS Ester
Cy5 Bis NHS Ester
Cy5 bis-NHS Ester
Cy5.5 Bis NHS Ester
Cy5.5 bis-NHS Ester
Cy7 Bis NHS Ester
Cy7 bis-NHS Ester
Unit Size:
10mg
50mg
5mg
Pack of 5
This item is not returnable. View return policy

Product Code. 11585045

Brand: Cytiva PA15004

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Suitable for general labeling of amine residues

Order Info

Large range of bulk packs available for greatest economy. The packs contain fluors carrying two reactive groups on each dye molecule for general labeling of protein amine groups.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Suitable for general labeling of Amine residues
Quantity 10 mg
Product Type Cy5 bis-NHS Ester
Kit Contents Foil packs, each containing 5, 10 or 50mg dried dye
Content And Storage 2°C to 8°C
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin sensitiser Category 1
  • Respiratory sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H317-May cause an allergic skin reaction.
  • H332-Harmful if inhaled.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Cy5 (Bis NHS ester)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.