Learn More
Cytiva Amersham™ CyDye™ Value Packs (bis-Reactive NHS Ester)
Suitable for general labeling of amine residues
Brand:  Cytiva PA17000
Code : GE
Additional Details : Weight : 0.02000kg
 View more versions of this product
Product Code. 11555055
  • £1095.00 / 5mg
Estimated Shipment: 10-07-2024
to see stock.
Add to basket
Specifications
Suitable for general labeling of Amine residues
Cy7 bis-NHS Ester
2°C to 8°C
5 mg
Foil packs, each containing 5, 10 or 50mg dried dye
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Signal Word
  • Warning
Hazard Category
  • Specific target organ toxicity Category 2