missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Amersham™ Cy™3, Cy™ 5 Maleimide mono-Reactive Dye 5-packs

Product Code. 10626315 Shop All Cytiva Products
Click to view available options
Label or Dye:
Cy3
Cy5
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 10626315

Brand: Cytiva PA23031

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

More selective method of antibody labeling that reduces the possibility of the label interfering with subsequent antibody-antigen reactions. Amersham™ Cy™3, Cy™ 5 Maleimide mono-Reactive Dye packs are packaged in premeasured amounts and foil-sealed to ensure consistent labelings.

Cy3 and Cy5 mono-functional maleimides are particularly suitable for the selective labeling of molecules containing free sulfhydryl groups, such as cysteine residues in proteins and peptides, and oligonucleotides. They are also useful in antibody labeling since the dye molecules are introduced away from the antibody binding site, increasing the chance that functionality of the labeled molecule will be unimpaired. The protocol is simple and robust, offering an alternative chemistry when either greater selectivity of labeling is required or amine labeling with Cy3 and Cy5 NHS esters is not desirable or appropriate.

  • Provides an alternative for when CyDye™ NHS esters are unsuitable.
  • Dyes are packaged in premeasured amounts and foil-sealed to ensure consistent labelings

Order Info

Each kit contains five vials of dye, each sufficient for labeling 1mg of protein.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) For the labelling of biological compounds with Cy3 monofunctional dye
Quantity 5 vials each sufficient to label 1 mg protein
Product Type Cy3 Maleimide Mono-Reactive Dye 5-Pack
Kit Contents Five vials of dye
Sufficient For Labeling 1mg Protein
Molecular Weight (g/mol) 791
Solubility Soluble in cold water and hot water
Content And Storage 2°C to 8°C, protected from light. Reconstituted material may be stored for up to 2 weeks at -20°C in aliquots to avoid repeat freeze thaw cycles.
Label or Dye Cy3
Color Red
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Skin sensitiser Category 1
  • Respiratory sensitiser Category 1
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.