missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ AMD 3465 hexahydrobromide
missing translation for 'orderingAttributeHoverText'
Quantity:
10 mg
50 mg
missing translation for 'unitSize'
10mg
50mg
Description
Potent, selective CXCR4 antagonist; exhibits 8-fold higher affinity than AMD 3100 (Cat.No. 3299). Inhibits SDF-1α-ligand binding (Ki = 41.7 nM). Potently inhibits HIV cell entry in vitro; causes leukocytosis and mobilizes haematopoietic stem cells in vivo.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 896.07 |
| Purity | >97% |
| Molecular Weight (g/mol) | 896.082 |
| Chemical Name or Material | AMD 3465 hexahydrobromide |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction