accessibility menu, dialog, popup

UserName

CAS RN 185991-07-5

HNHNNNHHNN

CAS RN 185991-07-5

IUPAC Name: [(pyridin-2-yl)methyl]({4-[(1,4,8,11-tetraazacyclotetradecan-1-yl)methyl]phenyl}methyl)amine
Synonyms: (pyridin-2-ylmethyl)({[4-(1,4,8,11-tetraazacyclotetradecan-1-ylmethyl)phenyl]methyl})amine
Molecular Weight (g/mol): 410.61
Molecular Formula: C24H38N6
InChi Key: CWJJHESJXJQCJA-UHFFFAOYSA-N
SMILES: C(NCC1=CC=CC=N1)C1=CC=C(CN2CCCNCCNCCCNCC2)C=C1
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Color 
  • (4)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results