missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMBRA1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | AMBRA1 |
|---|---|
| Concentration | 0.22 mg/mL |
| Applications | Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16832144
|
Proteintech
13762-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16894180
|
Proteintech
13762-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This antibody works well in WB, IHC and IF application.
AMBRA1 (activating molecule in Beclin-1-regulated autophagy protein 1) is a protein that bears a WD40 domain and regulates Beclin-1-dependent autophagy. In mouse embryos AMBRA1 has been shown to be critical for neural tube development.Specifications
| AMBRA1 | |
| Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse, Human, Rat | |
| A2AH22, Q9C0C7 | |
| 228361, 55626, 59319 | |
| AMBRA1 Fusion Protein Ag4735 | |
| Primary | |
| -20°C | |
| AMBRA1 |
| 0.22 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| AMBRA1, autophagy/beclin 1 regulator 1, KIAA1736, WDR94 | |
| Ambra1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title