missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMBRA1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13762-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody works well in WB, IHC and IF application.
AMBRA1 (activating molecule in Beclin-1-regulated autophagy protein 1) is a protein that bears a WD40 domain and regulates Beclin-1-dependent autophagy. In mouse embryos AMBRA1 has been shown to be critical for neural tube development.Specifications
| AMBRA1 | |
| Polyclonal | |
| Unconjugated | |
| AMBRA1 | |
| AMBRA1, autophagy/beclin 1 regulator 1, KIAA1736, WDR94 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 228361, 55626, 59319 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.22 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| A2AH22, Q9C0C7 | |
| Ambra1 | |
| AMBRA1 Fusion Protein Ag4735 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction