missing translation for 'onlineSavingsMsg'
Learn More

Duran™ Amber Narrow Neck Reagent Bottle

Brand:  Duran™ 211687305

Code : A0

Additional Details : Weight : 1.70000kg

 View more versions of this product

Product Code. 10078000

  • £284.00 / Each


Estimated Shipment
06-02-2018
Add to basket

Specifications

Specifications

5L
Amber
500°C
Bottle, Reagent
Yes
323mm
ISO 4796-2, USP Standard
Standard Ground Joint, Glass Flat-Head Stopper
1Pack
Yes
182mm
Narrow
Videos