Learn More
Fisherbrand™ Custom Cleaned Amber Boston Round Bottles
Product Code. 11744289 Shop All Fisherbrand Products
Click to view available options
Capacity (Metric):
60 mL
125 mL
250 mL
500 mL
1000 mL
Unit Size:
Pack of 12
Pack of 24
Product Code. 11744289
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Ideal storage for solvents, chemicals and samples. Fisherbrand™ Custom Cleaned Amber Boston Round Bottles are manufactured from amber colored soda glass.
  • Narrow mouth
  • Pre-cleaned for visible particulates using deionised water
  • With white polypropylene cap with PTFE faced foamed polyethylene liner
Specifications
Capacity (Metric) 60 mL
Capacity (English) 2.02 oz.
Cleaning Procedure USP purified Water
Color Amber
Material Glass
Quantity 24/PK.
Closure Color White
Liner Material PTFE
Closure Included Yes
Mouth Wide
Shape Round
Product Type Boston Round Bottle
Closure Liner Polyethylene
Closure Material PTFE
Closure Size 20-400 mm
Closure Type Screw Cap