missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Aluminum Nitride Cover for Crucible Stock

Product Code. 15487165
Click to view available options
Quantity:
10/Pk.
2/Pk.
Unit Size:
Pack of 10
Pack of 2
This item is not returnable. View return policy

Product Code. 15487165

Brand: Thermo Scientific™ 046313.KH

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Aluminum nitride ceramics exhibit exceptional characteristics, making them useful in various applications.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Unusually high thermal conductivity combined with good electrical insulation characteristics
  • Exceptional stability when exposed to many molten salts
  • Thermal stability up to at least 1800°C
  • Favorable mechanical characteristics extending into the high temperature range
  • Low thermal expansion and resistance to thermal shock
  • Special optical and acoustic characteristics.
Physical Properties
Flexural strength is 300 ± 5MPa
Coefficient of thermal expansion is 5.6x10-6K-1 (20-1000°C)
Thermal conductivity is 100-200 Wm-1K-1
Insulation resistance is >1012Ωcm (20°C)
TRUSTED_SUSTAINABILITY

Specifications

Description For crucible stock No. 43350
Material Aluminum Nitride
Quantity 2/Pk.
Product Type Aluminum Nitride Crucible Cover
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.