missing translation for 'onlineSavingsMsg'
Learn More

Tenak™ Aluminum Eco AluRack for Chest Freezers

Providing a whole new way of making storage racks. Tenak™ Aluminum Eco AluRacks for Chest Freezers fit into any freezer and can be used with all standard cryoboxes.

Brand:  Tenak™ TE11544

Additional Details : Weight : 1.60000kg

 View more versions of this product

Product Code. 12468140

  • £65.50 / Each
Estimated Shipment: 19-08-2024
to see stock.
Add to basket

Description

Description

  • Made of seawater resistant aluminum
  • Available in a range of box heights
  • Racks can be used with all freezer brands
  • Sold without cryoboxes
Specifications

Specifications

Eco AluRack
Aluminum
For chest freezers
1 Pack
140 mm
75 mm
Boxes (9)
Yes
1 Pack
140 x 140 x 707mm
707 mm
140 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers