missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Scaffolding Rods

Product Code. 11557023 Shop All Fisherbrand Products
Change view
Click to view available options
Length (Metric):
600 mm
900 mm
1200 mm
1800 mm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
11557023 600 mm Each
11577023 1200 mm Each
11587023 1800 mm Each
11567023 900 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11557023
Estimated Shipment: 14-10-2026
Add to basket
Add to basket

Provides for sturdy scaffolding construction in the laboratory. Fisherbrand™ Scaffolding Rods are manufactured from a durable aluminum alloy.

  • Approx. 13mm in diameter
  • Available in a range of lengths
  • Fits with Fisherbrand scaffolding connectors and foot adapters

Specifications

Material Aluminum Alloy
Length (Metric) 600 mm
Product Type Rod
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.