Learn More
Lovibond™ Aluminum #2 tablets
ECR method for comparators and photometers
£36.15
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
13161076
|
Lovibond™
515470BT |
100 Tablets |
£36.15
Pack of 100 |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
ECR method for comparators and photometers
Water analysis couldn't be easier. Precise dosing, long shelf life and easy handling make the tablet reagent the most popular indicator system and the cheapest way to have analyse a parameter.
The tablet reagent in the aluminum blister pack withstands almost all climatic conditions and external influences. In addition, there is no safety risk for the user.
Specifications
Aluminum #2 0.01 to 0.30 mg/L | |
Reagent |