Learn More
Lovibond™ Aluminum #2 tablets
Product Code. 13161076
Click to view available options
Quantity:
100 Tablets
Unit Size:
Pack of 100
Product Code. 13161076
Brand: Lovibond™ 515470BT
ECR method for comparators and photometers
Water analysis couldn't be easier. Precise dosing, long shelf life and easy handling make the tablet reagent the most popular indicator system and the cheapest way to have analyse a parameter.
The tablet reagent in the aluminum blister pack withstands almost all climatic conditions and external influences. In addition, there is no safety risk for the user.
Specifications
Quantity 100 Tablets
Range Aluminum #2 0.01 to 0.30 mg/L
Type Reagent
compliance-icons compliance-icons
Product Identifier
  • Aluminium No. 2
Signal Word
  • Warning
Hazard Category
  • Flammable solid Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H228-Flammable solid.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : methenamine