missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Haldenwanger™ Alumina Ceramic Tube Form Crucibles
Excellent for use in diverse, highly concentrated reagents both at boiling and room temperatures.
Brand: Haldenwanger™ TRGAL7 240 190 01000
Code : 02
Additional Details : Weight : 0.01000kg
Description
- For use in high-wear conditions such as mortars and pestles, ball mills, mouthpieces, shaft protection sleeves and more
- Alsint 99.7 aluminum ceramic
- Chemical resistance to a variety of acids and alkalis
Specifications
White | |
Alumina Ceramic | |
1 | |
100mm | |
Cylinder |
19mm | |
1750°C | |
24mm | |
Crucibles |