missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Standard SIL G Silica Layers on Alugram™ Aluminum Sheets UV254

Product Code. 12834031 Shop All Macherey-Nagel Products
Change view
Click to view available options
Dimensions (L x W):
10 x 20 cm
2.5 x 7.5 cm
20 x 20 cm
4 x 8 cm
5 x 10 cm
5 x 20 cm
5 x 7.5 cm
Unit Size:
Pack of 20
Pack of 200
Pack of 25
Pack of 50
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) unitSize
12834031 10 x 20 cm Pack of 20
12820951 2.5 x 7.5 cm Pack of 200
10517771 20 x 20 cm Pack of 25
10451193 4 x 8 cm Pack of 50
12860651 5 x 20 cm Pack of 50
10115001 5 x 7.5 cm Pack of 20
10756342 5 x 10 cm Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 12834031 Supplier Macherey-Nagel™ Supplier No. 818162
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Separate non-volatile mixtures with unmodified standard silica layers for TLC (Thin-Layer Chromatography). Standard SIL G Silica Layers on Alugram™ Aluminum Sheets UV254 come in a variety of sizes.

Separate non-volatile mixtures with unmodified standard silica layers for TLC (Thin-Layer Chromatography). Standard SIL G Silica Layers on Alugram™ Aluminum Sheets UV254 come in a variety of sizes.

Alugram™ Aluminum Sheets for TLC
  • Pre-coated ready to use plates
  • Broad range of plate sizes, surface coatings and backings
Absorbent
  • Silica 60
  • specific surface (BET) ∼ 500 m2/g
  • Mean pore size 60 Å
  • Specific pore volume 0.75 mL/g,
  • Particle size 5–17 μm
  • Standard grade
Thickness
  • For analytical plates 0.25 mm
  • For preparative plates 0.5 and 1 mm
  • For 2 mm preparative layers a slightly coarser material is used.
Inicators
  • Manganese activated zinc silicate with green fluorescence for short-wave UV (254 nm)
  • Special inorganic fluorescent pigment with blue fluorescence for long-wave UV (366 nm)
Binders
  • Highly polymeric products, which are stable in almost all organic solvents and resistant towards aggressive visualisation reagents

Specifications

Backing Material Alugram™ Aluminum
Particle Size 5 to 17 μm
Pore Size 60 Å
Quantity 20/Pk.
Type Aluminum Sheet
Fluorescent Indicator UV254
For Use With (Application) SIL G Plates
Pore Volume 0.75 mL/g
Sorbent Silica 60
Specific Surface ∼500 m2/g
Thickness (Metric) 0.20 mm
Dimensions (L x W) 10 x 20 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.