missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Standard SIL G Silica Layers on Alugram™ Aluminum Sheets

Separate non-volatile mixtures with unmodified standard silica layers for TLC (Thin-Layer Chromatography). Standard SIL G Silica Layers on Alugram™ Aluminum Sheets come in a variety of sizes.

Brand:  Macherey-Nagel™ 818161

Code : 16

Additional Details : Weight : 0.15300kg

 View more versions of this product

Product Code. 10377931

  • £52.20 / Pack of 50
Estimated Shipment: 10-05-2024
to see stock.
Add to basket

Description

Description

Separate non-volatile mixtures with unmodified standard silica layers for TLC (Thin-Layer Chromatography). Standard SIL G Silica Layers on Alugram™ Aluminum Sheets come in a variety of sizes.

Alugram™ Aluminum Sheets for TLC
  • Pre-coated ready to use plates
  • Broad range of plate sizes, surface coatings and backings
Absorbent
  • Silica 60
  • specific surface (BET) ∼ 500m2/g
  • Mean pore size 60 Å
  • Specific pore volume 0.75mL/g,
  • Particle size 5–17μm
  • Standard grade
Thickness
  • For analytical plates 0.25mm
  • For preparative plates 0.5 and 1mm
  • For 2mm preparative layers, a slightly coarser material is used
Inicators
  • Manganese activated zinc silicate with green fluorescence for short-wave UV (254nm)
  • Special inorganic fluorescent pigment with blue fluorescence for long-wave UV (366nm)
Binders
  • Highly polymeric products, which are stable in almost all organic solvents and resistant towards aggressive visualisation reagents
Specifications

Specifications

Aluminum Sheets Alugram SIL G
60 Å
TLC Plate
0.75 mL/g
0.2 mm
5 to 17 μm
50 Pk.
Thin Layer Chromatography (TLC)
∼500 m2/g
5 x 10 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers