missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALS2CR3 Antibody (S390-43), Novus Biologicals™
Mouse Monoclonal Antibody
£171.00 - £424.00
Specifications
| Antigen | ALS2CR3 |
|---|---|
| Clone | S390-43 |
| Dilution | Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
Description
ALS2CR3 Monoclonal specifically detects ALS2CR3 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
| ALS2CR3 | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
| Unconjugated | |
| Mouse | |
| Human, Rat | |
| 66008 | |
| Fusion protein amino acids 757-914 (C-terminus) of human TRAK2 | |
| Primary | |
| Detects 100kDa. Does not cross-react with TRAK1. |
| S390-43 | |
| Monoclonal | |
| Purified | |
| RUO | |
| ALS2CR3GRIF1, CALS-C, GRIF-1, KIAA0549amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 3, OIP98Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 3 protein, trafficking kinesin-binding protein 2, trafficking protein, kinesin binding 2 | |
| TRAK2 | |
| IgG2a | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title