missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALS2CR3 Antibody (S390-43), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59333
This item is not returnable.
View return policy
Description
ALS2CR3 Monoclonal specifically detects ALS2CR3 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| ALS2CR3 | |
| Monoclonal | |
| Unconjugated | |
| PBS pH7.4, 50% glycerol with 0.1% Sodium Azide | |
| TRAK2 | |
| Fusion protein amino acids 757-914 (C-terminus) of human TRAK2 | |
| 100 μg | |
| Primary | |
| Detects 100kDa. Does not cross-react with TRAK1. | |
| Store at -20C. | |
| IgG2a |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| S390-43 | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
| ALS2CR3GRIF1, CALS-C, GRIF-1, KIAA0549amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 3, OIP98Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 3 protein, trafficking kinesin-binding protein 2, trafficking protein, kinesin binding 2 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 66008 | |
| Human, Rat | |
| Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido