missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALS2CR12 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | ALS2CR12 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
ALS2CR12 Polyclonal specifically detects ALS2CR12 in Human samples. It is validated for Western Blot.Specifications
| ALS2CR12 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| 130540 | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 12 | |
| IgG | |
| 52 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title