Learn More
Ansell™ AlphaTec™ 5000 Ultrasonically Welded and Taped
Product Code. 17160905
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 10
Product Code. 17160905
Brand: Ansell™ OR50T0019806
Only null left
Add to basket
Add to basket
AlphaTec™ 5000 Ultrasonically welded and taped made of highly visible innovative material is strong, durable and suitable for workers in extremely hazardous areas, including HAZMAT response teams.
Made of highly visible innovative material is strong, durable and suitable for workers in extremely hazardous areas, including HAZMAT response teams.
  • Outstanding permeation performance to numerous organic and inorganic chemicals, biological hazards and chemical warfare agents
  • Exceptional barrier protection, providing andgt;480 minutes breakthrough time against 14 of 15 chemicals listed in EN ISO 6529
  • Highly visible bright orange colour for improved worker safety
  • Welded and taped seams, our highest barrier to liquids and particulates
  • Designed to protect, typical innovative features include dual zip systems
  • Multi-layer barrier material which is lightweight, yet strong and durable
  • Anti-static, tested according to EN 1149-5
TRUSTED_SUSTAINABILITY
Specifications
Product Type Chemical Protective Clothing
Size 2X-Large
Gender Unisex
Material Multi-layer Barrier Laminate Fabric
Color Orange
Disposable Reusable
Waist Style Elastic
Ankle Style Elastic, Boot Overflaps
Cuff Style Double, Elastic
Neck Style Collared
Hood Type Standard
Closure Type Double Zipper
Seam Type Welded, Taped
Sterility Non-sterile
Protection Class CE CATEGORY III
Resistance Chemicals, Anti-static, Particle
Certifications/Compliance (Europe) CE CATEGORY III (Type 3-B, Type 4-B, Type 5-B, EN 1073-2, EN 14126, EN 1149-5)
No. of Pockets None
For Use With (Application) Contaminated Environments, Chemical Handling, Filling, blending and charging of raw materials, Transferring liquids and solids, Opening and draining pumps valves or lines, Loading and unloading of proces equipment, Raw material sample collection, Concentration mixing and palletisation, Hazmat response, Assembly and disassembly of large pipes, Unexpected leakages spills or other releases