missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell™ AlphaTec™ 5000 Ultrasonically Welded and Taped
Description
Made of highly visible innovative material is strong, durable and suitable for workers in extremely hazardous areas, including HAZMAT response teams.
- Outstanding permeation performance to numerous organic and inorganic chemicals, biological hazards and chemical warfare agents
- Exceptional barrier protection, providing andgt;480 minutes breakthrough time against 14 of 15 chemicals listed in EN ISO 6529
- Highly visible bright orange colour for improved worker safety
- Welded and taped seams, our highest barrier to liquids and particulates
- Designed to protect, typical innovative features include dual zip systems
- Multi-layer barrier material which is lightweight, yet strong and durable
- Anti-static, tested according to EN 1149-5
Specifications
Specifications
| Product Type | Chemical Protective Clothing |
| Size | Large |
| Gender | Unisex |
| Material | Multi-layer Barrier Laminate Fabric |
| Color | Orange |
| Disposable | Reusable |
| Waist Style | Elastic |
| Ankle Style | Elastic, Boot Overflaps |
| Cuff Style | Double, Elastic |
| Neck Style | Collared |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?